Assay Details
| Gene Symbol: BCAT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331880.1 | NP_001320930.1 | 2 - 3 | 505 | 83 | |
| NM_001331881.1 | NP_001320929.1 | 2 - 3 | 505 | 83 | ||
| NM_001331882.1 | NP_001320928.1 | 2 - 3 | 468 | 83 | ||
| NM_001331883.1 | NP_001320927.1 | 2 - 3 | 339 | 83 | ||
| NM_100880.3 | NP_563859.1 | 2 - 3 | 339 | 83 | ||
| NM_179298.2 | NP_849629.1 | 2 - 3 | 339 | 83 | ||
| GenBank mRNA | AJ276123.1 | - | 2 - 3 | 219 | 83 | |
| AK316768.1 | - | 2 - 3 | 268 | 83 | ||
| AY054517.1 | - | 2 - 3 | 340 | 83 | ||
| AY128778.1 | - | 2 - 3 | 133 | 83 | ||
More Information
Set Membership: |
|

BCAT-1
Genomic Map