Assay Details
| Gene Symbol: BCAT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123785.1 | NP_001117257.1 | 5 - 6 | 1326 | 93 | |
| NM_001331880.1 | NP_001320930.1 | 7 - 8 | 1200 | 93 | ||
| NM_001331881.1 | NP_001320929.1 | 7 - 8 | 1200 | 93 | ||
| NM_001331882.1 | NP_001320928.1 | 7 - 8 | 1163 | 93 | ||
| NM_001331883.1 | NP_001320927.1 | 7 - 8 | 1034 | 93 | ||
| NM_100880.3 | NP_563859.1 | 7 - 8 | 1034 | 93 | ||
| NM_179298.2 | NP_849629.1 | 7 - 8 | 1034 | 93 | ||
| GenBank mRNA | AJ276123.1 | - | 7 - 8 | 914 | 93 | |
| AK316768.1 | - | 7 - 8 | 963 | 93 | ||
| AY054517.1 | - | 7 - 8 | 1035 | 93 | ||
| AY128778.1 | - | 7 - 8 | 828 | 93 | ||
More Information
Set Membership: |
|

BCAT-1
Genomic Map