Assay Details
| Gene Symbol: PDIL1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102024.4 | NP_173594.1 | 4 - 5 | 839 | 108 | |
| NM_179365.1 | NP_849696.1 | 4 - 5 | 748 | 108 | ||
| GenBank mRNA | AY035096.1 | - | 4 - 5 | 748 | 108 | |
| AY063059.1 | - | 4 - 5 | 682 | 108 | ||
More Information
Set Membership: |
|

PDIL1-1
Genomic Map