Assay Details
| Gene Symbol: PDIL1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102024.4 | NP_173594.1 | 8 - 9 | 1459 | 92 | |
| NM_179365.1 | NP_849696.1 | 8 - 9 | 1368 | 92 | ||
| GenBank mRNA | AY035096.1 | - | 8 - 9 | 1368 | 92 | |
| AY063059.1 | - | 8 - 9 | 1302 | 92 | ||
More Information
Set Membership: |
|

PDIL1-1
Genomic Map