Assay Details
Gene Symbol: RIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332758.1 | NP_001321074.1 | 2 - 3 | 379 | 71 | |
NM_102500.3 | NP_564281.2 | 1 - 2 | 467 | 71 | ||
NM_179382.1 | NP_849713.1 | 2 - 3 | 176 | 71 | ||
GenBank mRNA | AK117390.1 | - | 1 - 2 | 261 | 71 | |
BT024810.1 | - | 1 - 2 | 31 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
