Assay Details
| Gene Symbol: APT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198175.1 | NP_001185104.1 | 2 - 3 | 515 | 97 | |
| NM_001332762.1 | NP_001323015.1 | 2 - 3 | 175 | 97 | ||
| NM_102509.4 | NP_564284.1 | 2 - 3 | 497 | 97 | ||
| NM_179383.3 | NP_849714.1 | 1 - 2 | 682 | 97 | ||
| GenBank mRNA | AF325045.2 | - | 1 - 2 | 135 | 97 | |
| AY084300.1 | - | 2 - 3 | 385 | 97 | ||
| AY128377.1 | - | 1 - 2 | 129 | 97 | ||
| BT000370.1 | - | 1 - 2 | 84 | 97 | ||
| X58640.1 | - | 101 | 97 | |||
More Information
Set Membership: |
|

APT1
Genomic Map