Assay Details
| Gene Symbol: APT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332762.1 | NP_001323015.1 | 4 - 5 | 378 | 75 | |
| NM_102509.4 | NP_564284.1 | 4 - 5 | 700 | 75 | ||
| NM_179383.3 | NP_849714.1 | 3 - 4 | 885 | 75 | ||
| GenBank mRNA | AF325045.2 | - | 3 - 4 | 338 | 75 | |
| AY084300.1 | - | 4 - 5 | 588 | 75 | ||
| AY128377.1 | - | 3 - 4 | 332 | 75 | ||
| BT000370.1 | - | 3 - 4 | 287 | 75 | ||
| X58640.1 | - | 304 | 75 | |||
More Information
Set Membership: |
|

APT1
Genomic Map