Assay Details
| Gene Symbol: MMH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333547.1 | NP_001322993.1 | 7 - 8 | 825 | 79 | |
| NM_179467.3 | NP_849798.1 | 7 - 8 | 737 | 79 | ||
| GenBank mRNA | AF099971.1 | - | 7 - 8 | 712 | 79 | |
More Information
Set Membership: |
|

MMH-1
Genomic Map