Assay Details
| Gene Symbol: emb2742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112102.3 | NP_187873.1 | 11 - 12 | 907 | 101 | |
| GenBank mRNA | AY093110.1 | - | 11 - 12 | 818 | 101 | |
| BT008360.1 | - | 10 - 11 | 769 | 101 | ||
More Information
Set Membership: |
|

emb2742
Genomic Map