Assay Details
| Gene Symbol: HAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338016.1 | NP_001326496.1 | 12 - 13 | 4359 | 131 | |
| NM_112135.4 | NP_187904.1 | 12 - 13 | 4377 | 131 | ||
More Information
Set Membership: |
|

HAC5
Genomic Map