Assay Details
Gene Symbol: HAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338016.1 | NP_001326496.1 | 14 - 15 | 4579 | 91 | |
NM_112135.4 | NP_187904.1 | 14 - 15 | 4597 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
