Assay Details
| Gene Symbol: HAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338016.1 | NP_001326496.1 | 16 - 17 | 5165 | 73 | |
| NM_112135.4 | NP_187904.1 | 16 - 17 | 5183 | 73 | ||
More Information
Set Membership: |
|

HAC5
Genomic Map