Assay Details
| Gene Symbol: HAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338016.1 | NP_001326496.1 | 3 - 4 | 535 | 111 | |
| NM_112135.4 | NP_187904.1 | 3 - 4 | 553 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001338016.1 transcripts.
Set Membership: |
|

HAC5
Genomic Map