Assay Details
| Gene Symbol: DIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112191.3 | NP_187954.1 | 8 - 9 | 922 | 116 | |
| GenBank mRNA | AF145452.1 | - | 8 - 9 | 859 | 116 | |
| AK229269.1 | - | 8 - 9 | 920 | 116 | ||
| BT024889.1 | - | 8 - 9 | 781 | 116 | ||
More Information
Set Membership: |
|

DIN4
Genomic Map