Assay Details
| Gene Symbol: FUS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035614.2 | NP_001030691.1 | 1 - 2 | 314 | 106 | |
| NM_112201.5 | NP_566459.2 | 1 - 2 | 325 | 106 | ||
| GenBank mRNA | AK229428.1 | - | 1 - 2 | 252 | 106 | |
| AK317108.1 | - | 1 - 2 | 302 | 106 | ||
| AY034618.1 | - | 1 - 2 | 302 | 106 | ||
More Information
Set Membership: |
|

FUS9
Genomic Map