Assay Details
| Gene Symbol: FUS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035614.2 | NP_001030691.1 | 2 - 3 | 387 | 111 | |
| NM_112201.5 | NP_566459.2 | 2 - 3 | 398 | 111 | ||
| GenBank mRNA | AK229428.1 | - | 2 - 3 | 325 | 111 | |
| AK317108.1 | - | 2 - 3 | 375 | 111 | ||
| AY034618.1 | - | 2 - 3 | 375 | 111 | ||
More Information
Set Membership: |
|

FUS9
Genomic Map