Assay Details
Gene Symbol: BGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112225.5 | NP_187988.1 | 16 - 17 | 2116 | 94 | |
GenBank mRNA | AJ270297.1 | - | 16 - 17 | 2094 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
