Assay Details
Gene Symbol: BGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112225.5 | NP_187988.1 | 18 - 19 | 2656 | 60 | |
GenBank mRNA | AJ270297.1 | - | 18 - 19 | 2634 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
