Assay Details
Gene Symbol: BGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112225.5 | NP_187988.1 | 2 - 3 | 504 | 66 | |
GenBank mRNA | AJ270297.1 | - | 2 - 3 | 482 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
