Assay Details
| Gene Symbol: RHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112241.4 | NP_188003.1 | 10 - 11 | 928 | 94 | |
| NM_202579.1 | NP_974308.1 | 10 - 11 | 837 | 94 | ||
| GenBank mRNA | AY092988.1 | - | 10 - 11 | 772 | 94 | |
| AY128801.1 | - | 9 - 10 | 682 | 94 | ||
More Information
Set Membership: |
|

RHD3
Genomic Map