Assay Details
| Gene Symbol: RHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112241.4 | NP_188003.1 | 13 - 14 | 1250 | 89 | |
| NM_202579.1 | NP_974308.1 | 13 - 14 | 1159 | 89 | ||
| GenBank mRNA | AY092988.1 | - | 13 - 14 | 1094 | 89 | |
| AY128801.1 | - | 12 - 13 | 1004 | 89 | ||
More Information
Set Membership: |
|

RHD3
Genomic Map