Assay Details
| Gene Symbol: RHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112241.4 | NP_188003.1 | 8 - 9 | 811 | 113 | |
| NM_202579.1 | NP_974308.1 | 8 - 9 | 720 | 113 | ||
| GenBank mRNA | AY092988.1 | - | 8 - 9 | 655 | 113 | |
| AY128801.1 | - | 7 - 8 | 565 | 113 | ||
More Information
Set Membership: |
|

RHD3
Genomic Map