Assay Details
Gene Symbol: NF-YA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338098.1 | NP_001327488.1 | 1 - 2 | 593 | 115 | |
NM_112256.3 | NP_188018.1 | 2 - 3 | 946 | 115 | ||
GenBank mRNA | AB493613.1 | - | 1 - 2 | 282 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
