Assay Details
| Gene Symbol: NF-YA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338098.1 | NP_001327488.1 | 3 - 4 | 748 | 82 | |
| NM_112256.3 | NP_188018.1 | 4 - 5 | 1101 | 82 | ||
| GenBank mRNA | AB493613.1 | - | 3 - 4 | 437 | 82 | |
More Information
Set Membership: |
|

NF-YA6
Genomic Map