Assay Details
| Gene Symbol: ASIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112274.4 | NP_188034.1 | 1 | 1895 | 153 | |
| GenBank mRNA | AY072187.1 | - | 1 | 1567 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASIL2| Gene Symbol: ASIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112274.4 | NP_188034.1 | 1 | 1895 | 153 | |
| GenBank mRNA | AY072187.1 | - | 1 | 1567 | 153 | |
Set Membership: |
|