Assay Details
| Gene Symbol: FAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338116.1 | NP_001319548.1 | 6 - 7 | 3301 | 71 | |
| NM_112285.4 | NP_188044.1 | 6 - 7 | 3274 | 71 | ||
More Information
Set Membership: |
|

FAB1B
Genomic Map