Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342099.1 | NP_001328023.1 | 3 - 4 | 750 | 70 | |
NM_119310.4 | NP_567880.1 | 3 - 4 | 735 | 70 | ||
GenBank mRNA | BT003891.1 | - | 3 - 4 | 735 | 70 | |
BT005002.1 | - | 3 - 4 | 534 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
