Assay Details
Gene Symbol: ENP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342117.1 | NP_001327983.1 | 3 - 4 | 2844 | 85 | |
NM_119332.4 | NP_194910.2 | 3 - 4 | 2248 | 85 | ||
GenBank mRNA | AY150514.1 | - | 3 - 4 | 1249 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
