Assay Details
| Gene Symbol: PKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203955.1 | NP_001190884.1 | 9 - 10 | 980 | 137 | |
| NM_001342121.1 | NP_001328314.1 | 7 - 8 | 1054 | 137 | ||
| NM_119341.2 | NP_194918.2 | 7 - 8 | 1054 | 137 | ||
More Information
Set Membership: |
|

PKR2
Genomic Map