Assay Details
Gene Symbol: HSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342124.1 | NP_001329752.1 | 3 - 4 | 581 | 79 | |
NM_119353.4 | NP_194929.2 | 5 - 6 | 929 | 79 | ||
GenBank mRNA | AB206554.1 | - | 1 - 2 | 115 | 79 | |
AK228013.1 | - | 5 - 6 | 784 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
