Assay Details
Gene Symbol: HSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342124.1 | NP_001329752.1 | 7 - 8 | 1386 | 63 | |
NM_119353.4 | NP_194929.2 | 9 - 10 | 1746 | 63 | ||
GenBank mRNA | AB206554.1 | - | 5 - 6 | 932 | 63 | |
AK228013.1 | - | 9 - 10 | 1589 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
