Assay Details
| Gene Symbol: APX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119384.4 | NP_194958.2 | 4 - 5 | 535 | 122 | |
| GenBank mRNA | AK117784.1 | - | 4 - 5 | 494 | 122 | |
| BT008349.1 | - | 4 - 5 | 479 | 122 | ||
More Information
Set Membership: |
|

APX6
Genomic Map