Assay Details
| Gene Symbol: APX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119384.4 | NP_194958.2 | 7 - 8 | 752 | 70 | |
| GenBank mRNA | AK117784.1 | - | 7 - 8 | 711 | 70 | |
| BT008349.1 | - | 7 - 8 | 696 | 70 | ||
More Information
Set Membership: |
|

APX6
Genomic Map