Assay Details
| Gene Symbol: APX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119384.4 | NP_194958.2 | 9 - 10 | 943 | 75 | |
| GenBank mRNA | AK117784.1 | - | 9 - 10 | 902 | 75 | |
| BT008349.1 | - | 9 - 10 | 887 | 75 | ||
More Information
Set Membership: |
|

APX6
Genomic Map