Assay Details
| Gene Symbol: AT4G32370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119389.2 | NP_194963.2 | 5 - 6 | 773 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G32370| Gene Symbol: AT4G32370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119389.2 | NP_194963.2 | 5 - 6 | 773 | 154 | |
Set Membership: |
|