Assay Details
Gene Symbol: La1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036700.2 | NP_001031777.1 | 4 - 5 | 527 | 89 | |
NM_119425.5 | NP_567904.1 | 4 - 5 | 542 | 89 | ||
GenBank mRNA | AY056237.1 | - | 4 - 5 | 466 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
