Assay Details
| Gene Symbol: HB-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119441.5 | NP_195014.1 | 13 - 14 | 1977 | 79 | |
| GenBank mRNA | AJ441292.1 | - | 12 - 13 | 1566 | 79 | |
| AY099631.1 | - | 13 - 14 | 1952 | 79 | ||
| BT008798.1 | - | 12 - 13 | 1566 | 79 | ||
More Information
Set Membership: |
|

HB-8
Genomic Map