Assay Details
| Gene Symbol: HB-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119441.5 | NP_195014.1 | 8 - 9 | 1256 | 87 | |
| GenBank mRNA | AJ441292.1 | - | 7 - 8 | 845 | 87 | |
| AY099631.1 | - | 8 - 9 | 1231 | 87 | ||
| BT008798.1 | - | 7 - 8 | 845 | 87 | ||
More Information
Set Membership: |
|

HB-8
Genomic Map