Assay Details
Gene Symbol: HB-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119441.5 | NP_195014.1 | 9 - 10 | 1421 | 68 | |
GenBank mRNA | AJ441292.1 | - | 8 - 9 | 1010 | 68 | |
AY099631.1 | - | 9 - 10 | 1396 | 68 | ||
BT008798.1 | - | 8 - 9 | 1010 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
