Assay Details
| Gene Symbol: GLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125634.1 | NP_001119106.1 | 9 - 10 | 2783 | 94 | |
| NM_119455.3 | NP_195027.1 | 9 - 10 | 2873 | 94 | ||
| GenBank mRNA | AY063903.1 | - | 9 - 10 | 2751 | 94 | |
| AY091186.1 | - | 9 - 10 | 2601 | 94 | ||
More Information
Set Membership: |
|

GLDP1
Genomic Map