Assay Details
| Gene Symbol: ADR1-L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036704.2 | NP_001031781.1 | 2 - 3 | 1057 | 107 | |
| NM_001342211.1 | NP_001320123.1 | 1 - 2 | 1756 | 107 | ||
| GenBank mRNA | AK316970.1 | - | 1 - 2 | 689 | 107 | |
More Information
Set Membership: |
|

ADR1-L1
Genomic Map