Assay Details
| Gene Symbol: ADR1-L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036704.2 | NP_001031781.1 | 4 - 5 | 1799 | 85 | |
| NM_001342211.1 | NP_001320123.1 | 3 - 4 | 2498 | 85 | ||
| GenBank mRNA | AK316970.1 | - | 3 - 4 | 1431 | 85 | |
More Information
Set Membership: |
|

ADR1-L1
Genomic Map