Assay Details
| Gene Symbol: AT4G33440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119498.4 | NP_195070.2 | 1 - 2 | 904 | 56 | |
| GenBank mRNA | BT002785.1 | - | 1 - 2 | 503 | 56 | |
| BT004354.1 | - | 1 - 2 | 419 | 56 | ||
More Information
Set Membership: |
|

AT4G33440
Genomic Map