Assay Details
| Gene Symbol: AT4G33910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119550.3 | NP_567941.1 | 7 - 8 | 1139 | 83 | |
| GenBank mRNA | AY064069.1 | - | 7 - 8 | 924 | 83 | |
| AY086254.1 | - | 7 - 8 | 936 | 83 | ||
| AY096377.1 | - | 7 - 8 | 779 | 83 | ||
More Information
Set Membership: |
|

AT4G33910
Genomic Map