Assay Details
Gene Symbol: SSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160814.1 | NP_001154286.1 | 2 - 3 | 136 | 95 | |
NM_119552.3 | NP_567942.1 | 2 - 3 | 337 | 95 | ||
GenBank mRNA | BT026099.1 | - | 2 - 3 | 81 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
