Assay Details
Gene Symbol: OST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342244.1 | NP_001320129.1 | 6 - 7 | 1038 | 80 | |
NM_119556.3 | NP_567945.1 | 6 - 7 | 1210 | 80 | ||
GenBank mRNA | AY054624.1 | - | 6 - 7 | 636 | 80 | |
AY081538.1 | - | 6 - 7 | 591 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
