Assay Details
| Gene Symbol: LEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119573.5 | NP_567952.1 | 7 - 8 | 931 | 71 | |
| GenBank mRNA | AF360331.1 | - | 7 - 8 | 699 | 71 | |
| AY056339.1 | - | 7 - 8 | 644 | 71 | ||
More Information
Set Membership: |
|

LEJ1
Genomic Map