Assay Details
Gene Symbol: CAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119587.4 | NP_195149.1 | 4 - 5 | 1145 | 119 | |
GenBank mRNA | AY034919.1 | - | 4 - 5 | 924 | 119 | |
AY113919.1 | - | 4 - 5 | 871 | 119 | ||
AY302082.1 | - | 4 - 5 | 871 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
