Assay Details
| Gene Symbol: COW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342293.1 | NP_001328041.1 | 11 - 12 | 1717 | 77 | |
| NM_119624.3 | NP_195184.2 | 10 - 11 | 1605 | 77 | ||
More Information
Set Membership: |
|

COW1
Genomic Map