Assay Details
Gene Symbol: COW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342293.1 | NP_001328041.1 | 9 - 10 | 1206 | 100 | |
NM_119624.3 | NP_195184.2 | 8 - 9 | 1094 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
